Learn More
Medchemexpress LLC 2-(2,2,3,3,3-pentafluoropropoxy)-3-(4-(2,2,2-trifluoroethoxy)phenyl)-5,7-dihydropyrrolopyrimidine-4,6-dione | 1236767-85-3 | 99.9% | 473.27 | C17H11F8N3O4 | 100 MG
SDP
Catalog No. 5000226312
Encompass_Preferred
D5D-IN-326 is a selective, orally active delta-5 desaturase (D5D) inhibitor supplied as a powder for research use. It shows low-nanomolar potency (IC50 22 nM human, 72 nM rat), has molecular weight 473.27, and is reported at high purity (99.9%). Preclinical studies indicate improved insulin sensitivity and weight reduction in diet-induced obese mice. Store and handle per the manufacturer's recommendations.
  • Selective delta-5 desaturase inhibitor with low-nanomolar potency.
  • High purity suitable for pharmacology studies (99.9%).
  • Powder form, convenient for formulation and dosing.
  • Suitable for in vitro and in vivo pharmacology studies.
  • Certificate of analysis and safety data sheet available.
  • Defined storage conditions for powder and solutions to preserve stability.
Catalog No. 50-002-26312 Supplier Medchemexpress LLC Supplier No. HY117427100MG
D5D-IN-326 is a selective, orally active delta-5 desaturase (D5D) inhibitor supplied as a powder for research use. It shows low-nanomolar potency (IC50 22 nM human, 72 nM rat), has molecular weight 473.27, and is reported at high purity (99.9%). Preclinical studies indicate improved insulin sensitivity and weight reduction in diet-induced obese mice. Store and handle per the manufacturer's recommendations.
  • Selective delta-5 desaturase inhibitor with low-nanomolar potency.
  • High purity suitable for pharmacology studies (99.9%).
  • Powder form, convenient for formulation and dosing.
  • Suitable for in vitro and in vivo pharmacology studies.
  • Certificate of analysis and safety data sheet available.
  • Defined storage conditions for powder and solutions to preserve stability.