Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2,2,4,7-Tetramethyl-1,2,3,4-tetrahydroquinolin-1-yl)ethan-1-ol, 97%, Thermo Scientific™

Catalog No. BTB13839EE
Change view
Click to view available options
Quantity:
25 g
50 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BTB13839EE 50 g Amber Glass Bottle
BTB13839EB 25 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BTB13839EE Supplier Thermo Scientific Chemicals Supplier No. BTB13839EE

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 53817-44-0
Molecular Formula C15H23NO
Molecular Weight (g/mol) 233.355
InChI Key DICAWUKDDRKZDZ-UHFFFAOYSA-N
Synonym 2-2,2,4,7-tetramethyl-1,2,3,4-tetrahydroquinolin-1-yl ethan-1-ol, 1 2h-quinolineethanol, 3,4-dihydro-2,2,4,7-tetramethyl, 1-2-hydroxyethyl-1,2,3,4-tetrahydro-2,2,4,7-tetramethylquinoline, 2-2,2,4,7-tetramethyl-3,4-dihydroquinolin-1-yl ethanol, acmc-1amas, maybridge1_002615, 3,4-dihydro-2,2,4,7-tetramethyl-2h-quinoline-1-ethanol, 1 2h-quinolineethanol,3,4-dihydro-2,2,4,7-tetramethyl, 3,4-dihydro-2,2,4,7-tetramethyl-1 2h-quinolineethanol, 1,2,3,4-tetrahydro-2,2,4,7-tetramethyl-1-quinolineethanol
PubChem CID 103845
IUPAC Name 2-(2,2,4,7-tetramethyl-3,4-dihydroquinolin-1-yl)ethanol
SMILES CC1CC(N(C2=C1C=CC(=C2)C)CCO)(C)C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 50 g
Formula Weight 233.35
Percent Purity 97%
Chemical Name or Material 2-(2,2,4,7-Tetramethyl-1,2,3,4-tetrahydroquinolin-1-yl)ethan-1-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.