Learn More
Medchemexpress LLC 2-(2-(2-((6-chlorohexyl)oxy)ethoxy)ethoxy)ethan-1-ol | 1355955-95-1 | MFCD32705069 | 268.78 g·mol^-1 | C12H25ClO4 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW613607100MG
Encompass_Preferred
HO-PEG3-(CH2)6-Cl (2-(2-(2-((6-chlorohexyl)oxy)ethoxy)ethoxy)ethan-1-ol) is a hydroxy-terminated PEG3 linker bearing a 6-chlorohexyl reactive handle. It is used as a flexible, hydrophilic spacer in the synthesis of PROTACs and Halo-tag targeting conjugates, enabling attachment between target-binding ligands and recruitment motifs.
  • Hydroxy-terminated PEG3 spacer with a 6-chlorohexyl functional group.
  • Molecular formula C12H25ClO4; molecular weight 268.78 g·mol^-1.
  • Designed for use in PROTAC and Halo-tag conjugate synthesis.
  • Available in multiple pack sizes including 100 MG.
  • Analytical documentation available on request (CoA, H NMR, SDS).
Catalog No. 50-002-84730