Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(2-(2-((6-chlorohexyl)oxy)ethoxy)ethoxy)ethan-1-ol | 1355955-95-1 | MFCD32705069 | 95.0% | 268.78 g/mol | C12H25ClO4 | 250 MG
SDP

Catalog No. 5000284731
Encompass_Preferred

HO-PEG3-(CH2)6-Cl is a hydroxy-terminated PEG3 linker bearing a six-carbon chlorinated alkyl chain, used as a building block in PROTAC and bioconjugation chemistry. It serves as a flexible, hydrophilic spacer for attaching ligands, tags, or warheads during linker assembly and small-scale synthesis.

  • Polyethylene glycol (PEG3) spacer with terminal hydroxy and six-carbon chloride.
  • Designed for PROTAC linker assembly and bioconjugation.
  • Flexible and hydrophilic to improve solubility and spacing between functional groups.
  • Supplied in research-scale packs suitable for method development and small-scale synthesis.
  • Molecular formula C12H25ClO4; molecular weight 268.78 g/mol.
  • CAS number 1355955-95-1; supplier-reported purity typically 95%.

Catalog No. 50-002-84731 Supplier Medchemexpress LLC Supplier No. HYW613607250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HO-PEG3-(CH2)6-Cl is a hydroxy-terminated PEG3 linker bearing a six-carbon chlorinated alkyl chain, used as a building block in PROTAC and bioconjugation chemistry. It serves as a flexible, hydrophilic spacer for attaching ligands, tags, or warheads during linker assembly and small-scale synthesis.

  • Polyethylene glycol (PEG3) spacer with terminal hydroxy and six-carbon chloride.
  • Designed for PROTAC linker assembly and bioconjugation.
  • Flexible and hydrophilic to improve solubility and spacing between functional groups.
  • Supplied in research-scale packs suitable for method development and small-scale synthesis.
  • Molecular formula C12H25ClO4; molecular weight 268.78 g/mol.
  • CAS number 1355955-95-1; supplier-reported purity typically 95%.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.