Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-[2-(2-Aminoethoxy)ethoxy]acetic Acid 96.0+%, TCI America™
SDP

Catalog No. A3120250MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A3120250MG 250 mg
A31201G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A3120250MG Supplier TCI America Supplier No. A3120250MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 134978-97-5
Molecular Formula C6H13NO4
Molecular Weight (g/mol) 163.17
MDL Number MFCD13185897
InChI Key RUVRGYVESPRHSZ-UHFFFAOYSA-N
Synonym 8-Amino-3,6-dioxaoctanoic Acid, Amino-PEG2-acetic Acid
PubChem CID 362706
IUPAC Name 2-[2-(2-aminoethoxy)ethoxy]acetic acid
SMILES NCCOCCOCC(O)=O

Specifications

Quantity 250 mg
Formula Weight 163.17
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-[2-(2-Aminoethoxy)ethoxy]acetic Acid
TSCA No
Recommended Storage Freezer (-20 °C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.