Learn More
Medchemexpress LLC 2-(2-(2-bromoethoxy)ethoxy)ethyl methanesulfonate | 2702323-73-5 | 95.0% | C7H15BrO5S | 10MG
SDP
Catalog No. 5000260996
Encompass_Preferred
A PEG-based bifunctional linker with a bromide and a methanesulfonate functional group, designed for use in the synthesis and assembly of PROTAC molecules. It provides a short, flexible PEG3 spacer to connect ligands while enabling nucleophilic substitution chemistry.
  • Provides a short PEG3 spacer for flexibility and solubility.
  • Contains terminal bromide and methanesulfonate groups that act as leaving groups for conjugation.
  • Suitable for linker assembly and PROTAC synthesis via nucleophilic substitution.
  • High research-grade purity for consistent performance.
  • Available in small research sizes and supplied as a liquid for ease of handling.
Catalog No. 50-002-60996 Supplier Medchemexpress LLC Supplier No. HY13833610MG
A PEG-based bifunctional linker with a bromide and a methanesulfonate functional group, designed for use in the synthesis and assembly of PROTAC molecules. It provides a short, flexible PEG3 spacer to connect ligands while enabling nucleophilic substitution chemistry.
  • Provides a short PEG3 spacer for flexibility and solubility.
  • Contains terminal bromide and methanesulfonate groups that act as leaving groups for conjugation.
  • Suitable for linker assembly and PROTAC synthesis via nucleophilic substitution.
  • High research-grade purity for consistent performance.
  • Available in small research sizes and supplied as a liquid for ease of handling.