Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-[2-(2-methoxyethoxy)ethoxy]ethyl 4-methylbenzene-1-sulfonate | 62921-74-8 | MFCD06200731 | 98.8% | 318.39 g/mol | C14H22O6S | 500 MG
SDP

Catalog No. 5000210386
Encompass_Preferred

m-PEG3-Tos is a tosyl-terminated polyethylene glycol (PEG) linker used as a reactive PEG3 spacer for conjugation and PROTAC synthesis. The tosyl leaving group enables nucleophilic substitution reactions to attach functional groups or linker arms in medicinal chemistry and bioconjugation workflows.

  • Tosyl-terminated PEG3 linker for conjugation and PROTAC synthesis.
  • Chemical name: 2-[2-(2-methoxyethoxy)ethoxy]ethyl 4-methylbenzene-1-sulfonate.
  • CAS number: 62921-74-8.
  • Molecular formula: C14H22O6S.
  • Molecular weight: 318.39 g/mol.
  • Purity: 98.8%.
  • Package size: 500 MG.

Catalog No. 50-002-10386 Supplier Medchemexpress LLC Supplier No. HYW043277500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

m-PEG3-Tos is a tosyl-terminated polyethylene glycol (PEG) linker used as a reactive PEG3 spacer for conjugation and PROTAC synthesis. The tosyl leaving group enables nucleophilic substitution reactions to attach functional groups or linker arms in medicinal chemistry and bioconjugation workflows.

  • Tosyl-terminated PEG3 linker for conjugation and PROTAC synthesis.
  • Chemical name: 2-[2-(2-methoxyethoxy)ethoxy]ethyl 4-methylbenzene-1-sulfonate.
  • CAS number: 62921-74-8.
  • Molecular formula: C14H22O6S.
  • Molecular weight: 318.39 g/mol.
  • Purity: 98.8%.
  • Package size: 500 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.