Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,2,2-Tribromoethanol

Catalog No. T484025G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T484025G 5 g
T4840225G 25 g
T48402100G 100 g
T48402500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. T484025G Supplier Sigma Aldrich Supplier No. T484025G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2,2,2-Tribromoethanol

Specifications

CAS 75-80-9
Melting Point 73°C to 79°C (lit.)
Boiling Point 92°C to 93°C (10 mmHg)
Molecular Formula C2H3Br3O
Linear Formula Br3CCH2OH
MDL Number MFCD00004671
Quantity 5 g
Synonym Tribromoethyl alcohol
Molecular Weight (g/mol) 282.76
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.