Learn More
Selleck Chemical LLC 2,2,2-Tribromoethanol 50mg 75-80-9 TBE, Tribromoethanol
SDP
Supplier:  Selleck Chemical LLC S450850MG
Encompass_Preferred
2,2,2-Tribromoethanol (TBE, Tribromoethanol) is an organic compound used to anesthetize mice. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3902