Learn More
Apexbio Technology LLC 2,2,2-Tribromoethanol 75-80-9 500mg
SDP
Supplier:  Apexbio Technology LLC C6640500
Encompass_Preferred
2 2 2-Tribromoethanol is a small-molecule anesthetic agent that enhances gamma-aminobutyric acid (GABA) receptor activity thereby modulating central nervous system function It exerts its biological activity primarily through potentiation of GABAergic neurotransmission resulting in reversible sedation and anesthesia states Widely used in biomedical research 2 2 2-tribromoethanol facilitates controlled immobilization in rodent models during surgical procedures physiological measurements and behavioral analysis Based on these pharmacological properties 2 2 2-tribromoethanol serves as a valuable agent for experimental anesthesia and procedural support in laboratory rodent studies
Catalog No. 50-000-89844