Learn More
Apexbio Technology LLC 2,2,2-Trichloroethanol 115-20-8 1g
SDP
Catalog No. 5000090340
Encompass_Preferred
2 2 2-Trichloroethanol (CAS 115-20-8) is a biologically active small molecule recognized as the active metabolite of chloral hydrate It modulates neuronal electrophysiological properties by activating two-pore domain potassium (K2P) channels specifically TREK-1 and TRAAK Through this action 2 2 2-Trichloroethanol influences neuronal signaling pathways and membrane potential dynamics It is frequently utilized in experimental studies to investigate the molecular mechanisms underlying neurological disorders and to elucidate the pathophysiology of neural signaling alterations
Catalog No. 50-000-90340 Supplier Apexbio Technology LLC Supplier No. C68231000
2 2 2-Trichloroethanol (CAS 115-20-8) is a biologically active small molecule recognized as the active metabolite of chloral hydrate It modulates neuronal electrophysiological properties by activating two-pore domain potassium (K2P) channels specifically TREK-1 and TRAAK Through this action 2 2 2-Trichloroethanol influences neuronal signaling pathways and membrane potential dynamics It is frequently utilized in experimental studies to investigate the molecular mechanisms underlying neurological disorders and to elucidate the pathophysiology of neural signaling alterations