Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-2,2,2-Trifluoro-1-(9-anthryl)ethanol 98+%, Thermo Scientific™

Catalog No. AC199002500
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC199002500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC199002500 Supplier Thermo Scientific Chemicals Supplier No. 199002500

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53531-34-3
Molecular Formula C16H11F3O
Molecular Weight (g/mol) 276.26
MDL Number MFCD00001260,MFCD00062967
InChI Key ICZHJFWIOPYQCA-UHFFFAOYNA-N
Synonym r---2,2,2-trifluoro-1-9-anthryl ethanol, r---1-9-anthryl-2,2,2-trifluoroethanol, r-1-anthracen-9-yl-2,2,2-trifluoroethanol, 1r-1-9-anthracenyl-2,2,2-trifluoroethanol, 1r-1-9-anthryl-2,2,2-trifluoroethan-1-ol, r-+-2,2,2-trifluoro-1-9-anthryl ethanol, r---, a-trifluoromethyl anthracene-9-methanol, 1r-1-anthracen-9-yl-2,2,2-tris fluoranyl ethanol, r---alpha-trifluoromethyl-9-anthracenemethanol, 1r-1-anthracen-9-yl-2,2,2-trifluoroethanol
PubChem CID 7006444
IUPAC Name 1-(anthracen-9-yl)-2,2,2-trifluoroethan-1-ol
SMILES OC(C1=C2C=CC=CC2=CC2=CC=CC=C12)C(F)(F)F

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 132°C to 135°C
Color Yellow
Infrared Spectrum Authentic
Quantity 250 mg
Specific Rotation -29° to -31° (25°C, 589nm) (c=6, CHCl3)
Formula Weight 276.26
Percent Purity ≥98.0%
Physical Form Crystalline Powder
Chemical Name or Material (-)-2, 2, 2-Trifluoro-1-(9-anthryl)ethanol, 98+%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.