Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,2-Trifluoro-1-(9-anthryl)ethanol 99.0+%, TCI America™
SDP

Catalog No. T15221G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15221G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T15221G Supplier TCI America Supplier No. T15221G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 65487-67-4
Molecular Formula C16H11F3O
Molecular Weight (g/mol) 276.26
MDL Number MFCD00062967
InChI Key ICZHJFWIOPYQCA-UHFFFAOYNA-N
Synonym 1-(9-Anthryl)-2,2,2-trifluoroethanol, alpha-(Trifluoromethyl)-9-anthracenemethanol
PubChem CID 103802
IUPAC Name 1-(anthracen-9-yl)-2,2,2-trifluoroethan-1-ol
SMILES OC(C1=C2C=CC=CC2=CC2=CC=CC=C12)C(F)(F)F

Specifications

Melting Point 142°C
Quantity 1 g
Formula Weight 276.26
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2,2-Trifluoro-1-(9-anthryl)ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.