Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,2,2-trifluoro-N-[(1R,2S)-1-[1-(4-fluorophenyl)indazol-5-yl]oxy-1-(3-methoxyphenyl)propan-2-yl]acet | 1034148-04-3 | MFCD26142626 | 99.9% | 487.45 | C25H21F4N3O3 | 50 MG
SDP

Catalog No. 5000242592
Encompass_Preferred

AZD5423 is a non-steroidal, selective glucocorticoid receptor modulator provided as a research chemical for preclinical pharmacology and inhalation therapy studies. It is supplied for laboratory use only and has documented purity and storage recommendations.

  • Non-steroidal selective glucocorticoid receptor modulator for preclinical research.
  • High reported purity suitable for analytical and pharmacological studies.
  • Molecular formula C25H21F4N3O3 and molecular weight 487.45 g/mol.
  • Recommended storage conditions provided to maintain compound stability.
  • Available in multiple package sizes to support dose-ranging and formulation work.
  • Intended for laboratory research use only, not for clinical or human use.

Catalog No. 50-002-42592 Supplier Medchemexpress LLC Supplier No. HY10824350MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AZD5423 is a non-steroidal, selective glucocorticoid receptor modulator provided as a research chemical for preclinical pharmacology and inhalation therapy studies. It is supplied for laboratory use only and has documented purity and storage recommendations.

  • Non-steroidal selective glucocorticoid receptor modulator for preclinical research.
  • High reported purity suitable for analytical and pharmacological studies.
  • Molecular formula C25H21F4N3O3 and molecular weight 487.45 g/mol.
  • Recommended storage conditions provided to maintain compound stability.
  • Available in multiple package sizes to support dose-ranging and formulation work.
  • Intended for laboratory research use only, not for clinical or human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.