Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,2-Trifluoro-N-methoxy-N-methylacetamide 98.0+%, TCI America™
SDP

Catalog No. T33265G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T33265G 5 g
T332625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T33265G Supplier TCI America Supplier No. T33265G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 104863-67-4
Molecular Formula C4H6F3NO2
Molecular Weight (g/mol) 157.09
MDL Number MFCD00077565
InChI Key JDBJZZTZYKUFFK-UHFFFAOYSA-N
PubChem CID 2782382
IUPAC Name 2,2,2-trifluoro-N-methoxy-N-methylacetamide
SMILES CON(C)C(=O)C(F)(F)F

Specifications

Color Colorless
Boiling Point 52°C
Quantity 5 g
UN Number 1993
Formula Weight 157.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,2-Trifluoro-N-methoxy-N-methylacetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.