Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,2-Trifluoroacetamide 99+%, Thermo Scientific™

Catalog No. AC163102500
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC163102500 250 g
AC163100250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC163102500 Supplier Thermo Scientific Chemicals Supplier No. 163102500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 354-38-1
Molecular Formula C2H2F3NO
Molecular Weight (g/mol) 113.04
MDL Number MFCD00008008
InChI Key NRKYWOKHZRQRJR-UHFFFAOYSA-N
Synonym trifluoroacetamide, acetamide, 2,2,2-trifluoro, unii-36mu16byfk, n-trifluoroacetamide, 2,2,2-trifluoro-acetamide, 36mu16byfk, trifluroacetamide, trifluoro-acetamide, snylgqhjptadtaetp@, pubchem12622
PubChem CID 67717
IUPAC Name trifluoroacetamide
SMILES NC(=O)C(F)(F)F

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 68°C to 75°C
Color White to Yellow or Beige
Infrared Spectrum Authentic
Linear Formula CF3CONH2
Quantity 250 g
Beilstein 02, II, 186
Formula Weight 113.04
Percent Purity ≥99.0%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2, 2, 2-Trifluoroacetamide, 99+%
Show More Show Less
EINECSNumber 206-559-9
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.