Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,2,2-Trifluoroethanesulfonyl Chloride

Catalog No. 3247875G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-478-75G 5 g
32-478-71G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 32-478-75G Supplier Sigma Aldrich Supplier No. 3247875G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2,2,2-Trifluoroethanesulfonyl Chloride

Specifications

CAS 1648-99-3
Density 1.651 g/mL at 25°C
Boiling Point 140°C to 141°C; 64°C to 67°C (45 mmHg, lit.)
Molecular Formula C2H2ClF3O2S
Linear Formula CF3CH2SO2Cl
Refractive Index n20/D 1.388
MDL Number MFCD00007458
Quantity 5 g
Synonym Tresyl chloride
Molecular Weight (g/mol) 182.55 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.