Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,2-Trifluoroethanol, 99.8%, extra pure

Catalog No. AC139755000
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC139755000 500 mL Glass bottle
AC139750250 25 mL Glass bottle
AC139751000 100 mL Glass bottle
AC139750025 2.5 L Glass bottle
AC139750100 10 L Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. AC139755000 Supplier Thermo Scientific Chemicals Supplier No. 139755000
  • $451.50
    $383.65 / Each of 1

    Save $67.85

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 75-89-8 | C2H3F3O | 100.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-89-8
Molecular Formula C2H3F3O
Molecular Weight (g/mol) 100.04
MDL Number MFCD00004672
InChI Key RHQDFWAXVIIEBN-UHFFFAOYSA-N
Synonym trifluoroethanol, ethanol, 2,2,2-trifluoro, 2,2,2-trifluoroethan-1-ol, fluorinol 85, 2,2,2-trifluoroethyl alcohol, trifluoroethyl alcohol, trifluoro ethanol, tfe, perfluoro-1,1-dihydroethanol, tfetoh
PubChem CID 6409
ChEBI CHEBI:42330
IUPAC Name 2,2,2-trifluoroethanol
SMILES OCC(F)(F)F

Specifications

Melting Point -45.0°C
Color Colorless
Density 1.3900g/mL
pH 5.0 to 7.5 (10% v/v soln.)
Boiling Point 77.0°C to 80.0°C
Flash Point 29°C
Infrared Spectrum Authentic
Assay Percent Range 99.75% min. (GC)
Packaging Glass bottle
Refractive Index 1.2907
Linear Formula CF3CH2OH
Quantity 500 mL
Residue after Evaporation 0.005% max.
Specific Gravity 1.39
Merck Index 15,9848
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohols,ketons,esters and ethers
Viscosity 1.75 mPa.s (25°C)
Formula Weight 100.04
Percent Purity 99.8%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material 2, 2, 2-Trifluoroethanol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Causes serious eye damage.
Toxic if swallowed.
Toxic if inhaled.
May damage fertility.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:
EINECSNumber 200-913-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.