Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,2-Trifluoroethanol, ≥99.8% (GC), Eluent additive for LC-MS, Solstice

Catalog No. 60044574 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
50 mL
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-044-574 50 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-044-574 Supplier Solstice Supplier No. 0584150ML

missing translation for 'validMainframeMsg'

Eluent additive for LC-MS

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-89-8
Molecular Formula C2H3F3O
Molecular Weight (g/mol) 100.04
MDL Number MFCD00004672
InChI Key RHQDFWAXVIIEBN-UHFFFAOYSA-N
Synonym trifluoroethanol, ethanol, 2,2,2-trifluoro, 2,2,2-trifluoroethan-1-ol, fluorinol 85, 2,2,2-trifluoroethyl alcohol, trifluoroethyl alcohol, trifluoro ethanol, tfe, perfluoro-1,1-dihydroethanol, tfetoh
PubChem CID 6409
ChEBI CHEBI:42330
IUPAC Name 2,2,2-trifluoroethanol
SMILES OCC(F)(F)F

Specifications

Melting Point -44°C (literature)
Color Colorless
Density 1.391g/mL at 20°C, 1.373g/mL at 25°C
Boiling Point 77°C to 80°C
Flash Point 29°C (84.2°F)
Packaging Glass Bottle
Refractive Index n20/D 1.3(lit.)
Linear Formula CF3CH2OH
Quantity 50 mL
UN Number UN1986
Beilstein 1733203
Vapor Density 3.5 (vs air)
Vapor Pressure 70mmHg (25°C)
Formula Weight 100.04g/mol
Chemical Name or Material 2,2,2-Trifluoroethanol
Show More Show Less
Hazard Statement P261-P280-P301 + P310-P305 + P351 + P338-P311
Precautionary Statement H226-H301 + H331-H312-H315-H318-H335
DOTInformation 6.1(3)/PGIII
EINECSNumber 200-913-6
RTECSNumber KM5250000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.