Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,2-Trifluoroethanol-d{2}, 98% (Isotopic), Thermo Scientific Chemicals

Catalog No. AA4236203
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4236203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4236203 Supplier Thermo Scientific Chemicals Supplier No. 04236203
Add to Cart

missing translation for 'validMainframeMsg'

NMR Solvent

NMR Solvent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132248-58-9
Molecular Formula C2H3F3O
Molecular Weight (g/mol) 102.052
MDL Number MFCD00082609
InChI Key RHQDFWAXVIIEBN-DICFDUPASA-N
Synonym 2,2,2-trifluoroethanol-1,1-d2, trifluoroethanol-d2, 2,2,2-trifluoroethanol-d2, trifluoro 2 h? ethanol, 2,2,2-trifluoroethyl-1,1-d2 alcohol, 2 2 2-trifluoroethyl-1 1-d2 alcohol, 2,2,2-trifluoroethanol-1,1-d2 99.5 atom % d, 2,2,2-trifluoroethanol-1,1-d2, 99.5 atom % d, 2 2 2-trifluoroethyl-1 1-d2 alcohol 99.5
PubChem CID 12255976
IUPAC Name 1,1-dideuterio-2,2,2-trifluoroethanol
SMILES C(C(F)(F)F)O

Specifications

Physical Form Liquid
Quantity 1 g
Sensitivity Hygroscopic
Melting Point -44°C
Boiling Point 77°C to 80°C
UN Number UN1992
Formula Weight 102.02
Percent Purity 98%
Chemical Name or Material 2,2,2-Trifluoroethanol-d2, Isotopic
Hazard Category H226-H302+H312+H332-H315-H318-H335
Hazard Statement GHS H Statement
H331-H318-H226-H302-H312-H315-H335
Toxic if inhaled.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 200-913-6
TSCA No
Recommended Storage Ambient

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.