Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,2-Trifluoroethanol (Peptide Synthesis), Fisher BioReagents

Catalog No. BP622100 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP622-100 100 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP622-100

missing translation for 'validMainframeMsg'

Trifluoroethanol is often used in peptide synthesis procedures.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-89-8
Molecular Formula C2H3F3O
Molecular Weight (g/mol) 100.04
MDL Number MFCD00004672
InChI Key RHQDFWAXVIIEBN-UHFFFAOYSA-N
Synonym trifluoroethanol, ethanol, 2,2,2-trifluoro, 2,2,2-trifluoroethan-1-ol, fluorinol 85, 2,2,2-trifluoroethyl alcohol, trifluoroethyl alcohol, trifluoro ethanol, tfe, perfluoro-1,1-dihydroethanol, tfetoh
PubChem CID 6409
ChEBI CHEBI:42330
IUPAC Name 2,2,2-trifluoroethan-1-ol
SMILES OCC(F)(F)F

Specifications

Alpha Vector Trifluoroethanol
Boiling Range 72° to 76°C
Melting Point -45°C
Color Colorless
Density 1.390g/cm³
pH 5.0 to 7.5
Boiling Point 77°C
Assay Percent Range ≥99 %
Packaging Glass Bottle
Linear Formula CF3CH2OH
Quantity 100 mL
Merck Index 15, 9848
Vapor Density 3.45g/mol
Vapor Pressure 70mmHg at 25°C
Formula Weight 100.04
Percent Purity ≥99%
Grade Peptide Synthesis
Physical Form Liquid
Chemical Name or Material 2, 2, 2-Trifluoroethanol
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Recommended Storage RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.