Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,2,2-Trifluoroethyl methacrylate

Catalog No. 37376125G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-376-125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 37-376-125G Supplier Sigma Aldrich Supplier No. 37376125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2,2,2-Trifluoroethyl methacrylate (TFEMA) is a semifluorinated monomer that can be synthesized from methacryloyl chloride and 2,2,2-trifluoroethanol in presence of triethylamine. Its properties include chemical inertness, good wear resistance and low dielectric constant.TFEMA can be used in the preparation of poly(TFEMA), which can be used in the development of acrylic protective coatings.

Specifications

CAS 352-87-4
Density 1.181 g/mL (at 25°C)
Boiling Point 59°C (100 mmHg)
Molecular Formula C6H7F3O2
Refractive Index n20/D 1.361 (literature)
Linear Formula H2C=C(CH3)CO2CH2CF3
Quantity 25 g
MDL Number MFCD00013576
Synonym 2,2,2-Trifluoroethyl 2-methylprop-2-enoate
Molecular Weight (g/mol) 168.11
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.