Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,2,2-Trifluoroethyl Trifluoromethanesulfonate

Catalog No. 7529245G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
75-292-45G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 75-292-45G Supplier Sigma Aldrich Supplier No. 7529245G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reagent used in thenantioselective preparation of cyclic N-aryl hydroxamic acids via phase-transfer catalyzed alkylation of nitrobenzyl bromides to give nirophenylalanines

Specifications

CAS 757958-40-0
Melting Point 76°C to 79°C
Molecular Formula C18H26BrP
Refractive Index n20/D 1.306
Linear Formula C18H26BrP
MDL Number MFCD21363039
Quantity 1 g
Synonym Trifluoromethanesulfonic acid 2,2,2-trifluoroethyl ester
Molecular Weight (g/mol) 353.28 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.