Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,2,3,3,3-Pentafluoro-1-propanol

Catalog No. 25747825G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-747-825G 25 g
25-747-85G 5 g
25-747-8250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 25-747-825G Supplier Sigma Aldrich Supplier No. 25747825G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2,2,3,3,3-pentafluoro-1-propanol is an alternative cleaning agent for chlorofluorocarbon.2,2,3,3,3-pentafluoro-1-propanol has been used:
  • As derivatization reagent in detection of unlabeled and 15n2 -labeled L-tryptophan, L-kynurenine, serotonin and quinolinic acid in human and rat plasma by GC-ms
  • As derivatization reagent in simultaneous analysis of cocaine, cocaethylene and their possible metabolic and pyrolytic products in blood, urine and muscle by GC-ms
  • Preparation of trifluoromethyl ynamines which, in turn, converted aldehydes to α-trifluoromethyl-α,α-unsaturated amides
  • To generate fluorinated α-keto ethers with alkenes and has wide applications in the expanding fluorous research area

Specifications

CAS 422-05-9
Density 1.505 g/mL (at 25°C (literature))
Boiling Point 80°C (748 mmHg)
Molecular Formula C3H3F5O
Refractive Index n20/D 1.288 (literature)
Linear Formula CF3CF2CH2OH
MDL Number MFCD00004673
Quantity 25 g
Synonym PFPOH
Molecular Weight (g/mol) 150.05
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.