Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,2,3,3,4,4,4-Heptafluoro-1-butanol

Catalog No. H16045G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H16045G 5 g
H160425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H16045G Supplier Sigma Aldrich Supplier No. H16045G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2,2,3,3,4,4,4-Heptafluoro-1-butanol can be used to synthesize poly(2-hydroxyethyl vinyl ether)-block-poly(2-(2,2,3,3,4,4,4-heptafluorobutoxy)ethyl vinyl ether) (poly(HOVE-b-HFBOVE), a fluorinated amphiphilic block copolymer.

Specifications

CAS 375-01-9
Density 1.6 g/mL (at 25°C (literature))
Boiling Point 96°C to 97°C (lit.)
Molecular Formula C4H3F7O
Refractive Index n20/D 1.3 (literature)
Linear Formula CF3CF2CF2CH2OH
MDL Number MFCD00004674
Quantity 5 g
Synonym Perfluoropropyl carbinol
Molecular Weight (g/mol) 200.05
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.