Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,3,3,4,4,4-Heptafluorobutyl acrylate, 97%, stab. with 100 ppm 4-methoxyphenol

Catalog No. AAL1040014
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1040014 25 g
AAL1040006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1040014 Supplier Thermo Scientific Chemicals Supplier No. L1040014
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 424-64-6 | C7H5F7O2 | 254.104 g/mol

2,2,3,3,4,4,4-Heptafluorobutyl acrylate is a useful fluorinated acrylate for proteomics research. It is also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,3,3,4,4,4-Heptafluorobutyl acrylate is a useful fluorinated acrylate for proteomics research. It is also used as pharmaceutical intermediate.

Solubility
Not miscible in water.

Notes
Light sensitive. Incompatible with oxidizing agents, heat, ultraviolet radiation and free radical initiators.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 424-64-6
Molecular Formula C7H5F7O2
Molecular Weight (g/mol) 254.104
MDL Number MFCD00039252
InChI Key PLXOUIVCSUBZIX-UHFFFAOYSA-N
Synonym 1h,1h-heptafluorobutyl acrylate, 2,2,3,3,4,4,4-heptafluorobutyl acrylate, heptafluorobutyl acrylate, 2-propenoic acid, 2,2,3,3,4,4,4-heptafluorobutyl ester, 2,2,3,3,4,4,4-heptafluorobutyl acrylate, stab. with 100 ppm 4-methoxyphenol, 1h, 1h-heptafluorobutylacrylate, 1,1-dihydroheptafluorobutyl acrylate, 1h,1h-heptafluorobut-1-yl acrylat, 2,2,3,3,4,4,4-heptafluorobutyl=propenoate
PubChem CID 67923
IUPAC Name 2,2,3,3,4,4,4-heptafluorobutyl prop-2-enoate
SMILES C=CC(=O)OCC(C(C(F)(F)F)(F)F)(F)F

Specifications

Density 1.418
Boiling Point 57°C to 58°C (30 mmHg)
Flash Point 31°C (87°F)
Odor Pungent
Refractive Index 1.331
Quantity 25 g
UN Number UN3272
Beilstein 1792520
Solubility Information Not miscible in water.
Formula Weight 254.11
Percent Purity 97%
Chemical Name or Material 2,2,3,3,4,4,4-Heptafluorobutyl acrylate, stabilized with 100ppm 4-methoxyphenol
Show More Show Less
Hazard Category H226-H302-H312-H315-H319-H332-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 207-036-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.