Learn More
Cambridge Isotope Laboratories 2 2 3 3 4 4 5 5-OctaCB (PCB-194) (13C12 98%) 40 /-2 ug/mL in nonane 3 mL
Catalog No. 5000167042
Encompass
2 2 3 3 4 4 5 5-OctaCB (PCB-194) (13C12 98%) 40 /-2 ug/mL in nonane 3 mL
Catalog No. 50-001-67042 Supplier Cambridge Isotope Laboratories Supplier No. EC14183
2 2 3 3 4 4 5 5-OctaCB (PCB-194) (13C12 98%) 40 /-2 ug/mL in nonane 3 mL