Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,3,3,4,4-Hexafluoropentanedioic Anhydride 97.0+%, TCI America™
SDP

Catalog No. H07455G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H07455G 5 g
H074525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H07455G Supplier TCI America Supplier No. H07455G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 376-68-1
Molecular Formula C5F6O3
Molecular Weight (g/mol) 222.042
MDL Number MFCD00006680
InChI Key IHYAGCYJVNHXCT-UHFFFAOYSA-N
Synonym hexafluoroglutaric anhydride, perfluoroglutaric anhydride, 3,3,4,4,5,5-hexafluorodihydro-2h-pyran-2,6 3h-dione, hexafluoroglutaric acid anhydride, 2,2,3,3,4,4-hexafluoropentanedioic anhydride, 3,3,4,4,5,5-hexafluoro-3h-4,5-dihydropyran-2,6-dione, hexafluoroglutaricanhydride, acmc-1clz4, hexafluorooxane-2,6-dione
PubChem CID 78995
IUPAC Name 3,3,4,4,5,5-hexafluorooxane-2,6-dione
SMILES C1(=O)C(C(C(C(=O)O1)(F)F)(F)F)(F)F

Specifications

Color Yellow
Boiling Point 72°C
Quantity 5 g
Formula Weight 222.04
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material 2,2,3,3,4,4-Hexafluoropentanedioic Anhydride
RTECSNumber TM0860000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.