Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,3,3-d(4)-3-(Trimethylsilyl)propionic acid sodium salt, 98+ atom % D

Catalog No. AAA1448903
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1448903 1 g
AAA14489MD 250 mg
AAA1448906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1448903 Supplier Thermo Scientific Chemicals Supplier No. A1448903
  • $242.00
    $192.65 / Each of 1

    Save $49.35

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 24493-21-8 | C6H13NaO2Si | 172.267 g/mol

2,2,3,3-d(4)-3-(Trimethylsilyl)propionic acid sodium salt is used as a deuterium labeled silane. Also used in pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,3,3-d(4)-3-(Trimethylsilyl)propionic acid sodium salt is used as a deuterium labeled silane. Also used in pharmaceutical industry.

Solubility
Soluble in water.

Notes
It is hygroscopic. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24493-21-8
Molecular Formula C6H13NaO2Si
Molecular Weight (g/mol) 172.267
MDL Number MFCD00002762
InChI Key OIIWPAYIXDCDNL-HGFPCDIYSA-M
Synonym sodium 3-trimethylsilyl 2,2,3,3-2h4 propionate, 3-trimethylsilyl propionic acid-d4 sodium salt, 2,2,3,3-d4-3-trimethylsilyl propionic acid sodium salt, sodium 3-trimethylsilyl 2 h? propanoate, sodium 3-trimethylsilyl 2h4 propanoate, tsp-d4, propanoic-2,2,3,3-d4 acid, 3-trimethylsilyl-, sodium salt, sodium 3-trimethylsilyl propionate-2,2,3,3-d4, 3-trimethylsilyl propionic-2,2,3,3-d4 acid sodium salt, sodium-3-trimethylsilyl-2,2,3,3-tetradeuteriopropionate
PubChem CID 23688921
IUPAC Name sodium;2,2,3,3-tetradeuterio-3-trimethylsilylpropanoate
SMILES C[Si](C)(C)CCC(=O)[O-].[Na+]

Specifications

Melting Point >300°C
Linear Formula (CH3)3SiCD2CD2CO2Na
Quantity 1 g
Beilstein 4046239
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 172.27
Assay ≥98 atom %D
Chemical Name or Material 2,2,3,3-d(4)-3-(Trimethylsilyl)propionic acid sodium salt
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 246-286-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.