Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,3,3-Tetrafluorocyclobutanol, 95%

Catalog No. AAL1918603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1918603 1 g
AAL1918606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1918603 Supplier Thermo Scientific Chemicals Supplier No. L1918603
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 374-32-3 | C4H4F4O | 144.069 g/mol

2,2,3,3-Tetrafluorocyclobutanol is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,3,3-Tetrafluorocyclobutanol is used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 374-32-3
Molecular Formula C4H4F4O
Molecular Weight (g/mol) 144.069
MDL Number MFCD00190085
InChI Key FNENUTDUAXPVTF-UHFFFAOYSA-N
Synonym 2,2,3,3-tetrafluorocyclobutanol, cyclobutanol, 2,2,3,3-tetrafluoro, cyclobutanol,2,2,3,3-tetrafluoro
PubChem CID 2776621
IUPAC Name 2,2,3,3-tetrafluorocyclobutan-1-ol
SMILES C1C(C(C1(F)F)(F)F)O

Specifications

Density 1.414
Boiling Point 155°C to 157°C
Flash Point 44°C (111°F)
Refractive Index 1.368
Quantity 1 g
UN Number UN1987
Beilstein 2429286
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 144.07
Percent Purity 95%
Chemical Name or Material 2,2,3,3-Tetrafluorocyclobutanol
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
TSCA No

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.