Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,3,3-Tetrafluoropropionic Acid 95.0+%, TCI America™
SDP

Catalog No. T17775G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T17775G Supplier TCI America Supplier No. T17775G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 756-09-2
Molecular Formula C3H2F4O2
Molecular Weight (g/mol) 146.041
MDL Number MFCD00054686
InChI Key PXRROZVNOOEPPZ-UHFFFAOYSA-N
Synonym flupropanate, 3h-tetrafluoropropionic acid, 2,2,3,3-tetrafluoropropionic acid, unii-376ks3j2q6, propanoic acid, 2,2,3,3-tetrafluoro, flupropanate iso, tetrafluoropropionic acid, pxrrozvnooeppz-uhfffaoysa
PubChem CID 69789
ChEBI CHEBI:82013
IUPAC Name 2,2,3,3-tetrafluoropropanoic acid
SMILES C(C(C(=O)O)(F)F)(F)F

Specifications

Color Yellow
Boiling Point 133°C
Quantity 5 g
UN Number 3265
Formula Weight 146.04
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,2,3,3-Tetrafluoropropionic Acid
EINECSNumber (2)-1190
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.