Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,3,3-Tetrafluoropropyl Methacrylate 98.0+%, TCI America™
SDP

Catalog No. M115025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M115025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M115025G Supplier TCI America Supplier No. M115025G

Chemical Identifiers

CAS 45102-52-1
Molecular Formula C7H8F4O2
Molecular Weight (g/mol) 200.133
MDL Number MFCD00042381
InChI Key RSVZYSKAPMBSMY-UHFFFAOYSA-N
Synonym 2,2,3,3-tetrafluoropropyl methacrylate, 2-propenoic acid, 2-methyl-, 2,2,3,3-tetrafluoropropyl ester, methacrylic acid 2,2,3,3-tetrafluoropropyl ester, 1h,1h,3h-tetrafluoropropyl methacrylate, 2,2,3,3-tetrafluoropropyl methacrylate, stab. with 2,2'-methylenebis 6-tert-butyl-4-methyl-phenol, acmc-1ajz7, ksc489s8d, rsvzyskapmbsmy-uhfffaoysa, 2,2,3,3-tetrafluoroprop-1-yl methacrylat
PubChem CID 123515
IUPAC Name 2,2,3,3-tetrafluoropropyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCC(C(F)F)(F)F

Specifications

Color Colorless
Quantity 25 g
UN Number 3272
Formula Weight 200.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,3,3-Tetrafluoropropyl Methacrylate [stabilized with 2,2′-Methylenebis(6-tert-butyl-4-methy
EINECSNumber (2)-1031
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.