Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2',3,4,4',5'-Hexachlorobiphenyl Solution, 100μg/mL in Hexane, Ultra Scientific

Catalog No. USRPC088S
Encompass
Change view
Click to view available options
Quantity:
1 x 2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-RPC-088S 1 x 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. US-RPC-088S Supplier Agilent Technologies Supplier No. RPC088S
May include imposed supplier surcharges.
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 35065-28-2 , 110-54-3
Molecular Formula C12H4Cl6
Molecular Weight (g/mol) 360.864
InChI Key RPUMZMSNLZHIGZ-UHFFFAOYSA-N
PubChem CID 37035
IUPAC Name 1,2,3-trichloro-4-(2,4,5-trichlorophenyl)benzene
SMILES C1=CC(=C(C(=C1C2=CC(=C(C=C2Cl)Cl)Cl)Cl)Cl)Cl

Specifications

Physical Form Liquid
Quantity 1 x 2 mL
Concentration or Composition (by Analyte or Components) 100μg/mL
Chemical Name or Material 2,2',3,4,4',5'-Hexachlorobiphenyl Solution, 100μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.