Learn More
Medchemexpress LLC 2-(2,3-dihydro-1,3,6-trimethyl-2-oxo-1H-benzimidazol-5-yl)-6-(3-hydroxypropyl)-1H-benz[de]i | 2080306-23-4 | ≥98.0% | 429.5 Da | C25H23N3O4 | 100 MG
SDP
Catalog No. 5000241410
Encompass_Preferred
BAY-299 is a small-molecule chemical probe that potently inhibits bromodomain-containing proteins, primarily BRPF2 (BRD1) and TAF1 family bromodomains. It is intended for research use in biochemical and cellular assays to probe bromodomain function, validate targets, and investigate chromatin-mediated transcriptional regulation. The compound is supplied as a solid with high purity suitable for laboratory studies.
  • Potent biochemical inhibition of BRPF2 and TAF1 family bromodomains.
  • Reported IC50 values: BRPF2 BD - 67 nM; TAF1 BD2 - 8 nM; TAF1L BD2 - 106 nM.
  • Suitable for target validation and mechanistic studies in epigenetics.
  • High purity (≥98%) for reliable assay performance.
  • Molecular weight 429.5 Da and chemical formula C25H23N3O4.
  • Supplied as a solid for routine laboratory handling.
Catalog No. 50-002-41410 Supplier Medchemexpress LLC Supplier No. HY107424100MG
BAY-299 is a small-molecule chemical probe that potently inhibits bromodomain-containing proteins, primarily BRPF2 (BRD1) and TAF1 family bromodomains. It is intended for research use in biochemical and cellular assays to probe bromodomain function, validate targets, and investigate chromatin-mediated transcriptional regulation. The compound is supplied as a solid with high purity suitable for laboratory studies.
  • Potent biochemical inhibition of BRPF2 and TAF1 family bromodomains.
  • Reported IC50 values: BRPF2 BD - 67 nM; TAF1 BD2 - 8 nM; TAF1L BD2 - 106 nM.
  • Suitable for target validation and mechanistic studies in epigenetics.
  • High purity (≥98%) for reliable assay performance.
  • Molecular weight 429.5 Da and chemical formula C25H23N3O4.
  • Supplied as a solid for routine laboratory handling.