Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(2,3-dihydro-1,3,6-trimethyl-2-oxo-1H-benzimidazol-5-yl)-6-(3-hydroxypropyl)-benz[de]isoquinoline | 2080306-23-4 | MFCD30480933 | 99.4% | 429.47 g·mol⁻¹ | C25H23N3O4 | 1 ML
SDP

Catalog No. 5000241016
Encompass_Preferred

BAY-299 is a small-molecule chemical probe that selectively targets bromodomain-containing proteins, inhibiting BRPF2 (BRD1) and the second bromodomains of TAF1 and TAF1L. It is used in biochemical and cell-based research to probe bromodomain function and epigenetic regulation.

  • Potent dual inhibitor with reported IC50s: BRPF2 BD 67 nM, TAF1 BD2 8 nM, TAF1L BD2 106 nM.
  • High purity as supplied: 99.4% (HPLC).
  • Chemical formula C25H23N3O4 and molecular weight 429.47 g·mol⁻¹.
  • Available as a 10 mM solution in DMSO (1 mL) and as solid quantities from 5 mg to 500 mg.
  • Supplied with SMILES and structure information for computational and cheminformatics use.

Catalog No. 50-002-41016 Supplier Medchemexpress LLC Supplier No. HY10742410MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BAY-299 is a small-molecule chemical probe that selectively targets bromodomain-containing proteins, inhibiting BRPF2 (BRD1) and the second bromodomains of TAF1 and TAF1L. It is used in biochemical and cell-based research to probe bromodomain function and epigenetic regulation.

  • Potent dual inhibitor with reported IC50s: BRPF2 BD 67 nM, TAF1 BD2 8 nM, TAF1L BD2 106 nM.
  • High purity as supplied: 99.4% (HPLC).
  • Chemical formula C25H23N3O4 and molecular weight 429.47 g·mol⁻¹.
  • Available as a 10 mM solution in DMSO (1 mL) and as solid quantities from 5 mg to 500 mg.
  • Supplied with SMILES and structure information for computational and cheminformatics use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.