Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(2,3-dioxo-1,2,3,4-tetrahydroquinoxaline-6-sulfonamido) propanoic acid | 112170-24-8 | MFCD05148320 | 98.9% | 313.29 | C11H11N3O6S | 25 MG
SDP

Catalog No. 5000307508
Encompass_Preferred

BCI-137 is a competitive small-molecule inhibitor of Argonaute 2 (AGO2) that binds residues in the MID domain and inhibits AGO2 activity (reported IC50 342 μM). It is supplied as a white to off-white solid intended for research use.

  • Competitive AGO2 inhibitor; IC50 342 μM.
  • High purity solid (listed purity 98.88%).
  • Molecular formula C11H11N3O6S; molecular weight 313.29 g·mol⁻¹.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Supplied as a 25 mg package for laboratory research.

Catalog No. 50-003-07508 Supplier Medchemexpress LLC Supplier No. HY11902425MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BCI-137 is a competitive small-molecule inhibitor of Argonaute 2 (AGO2) that binds residues in the MID domain and inhibits AGO2 activity (reported IC50 342 μM). It is supplied as a white to off-white solid intended for research use.

  • Competitive AGO2 inhibitor; IC50 342 μM.
  • High purity solid (listed purity 98.88%).
  • Molecular formula C11H11N3O6S; molecular weight 313.29 g·mol⁻¹.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Supplied as a 25 mg package for laboratory research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.