Learn More
Medchemexpress LLC 2-[2-[[(3R)-1-[8-[(4-tert-butyl-1,3-thiazol-2-yl)carbamoyl]-4-oxo-3-[(E)-2-(2H-tetrazol-5-yl)ethenyl]pyrido... | 957471-96-4 | 99.9% | 693.78 | C31H39N11O6S | 25MG
SDP
Catalog No. 5000239474
Encompass_Preferred
D13-9001 is a small-molecule inhibitor of RND-family bacterial efflux pump subunits AcrB (AcrAB-TolC) and MexB (MexAB-OprM). It shows binding affinities (KD ≈ 1.15 μM for AcrB and 3.57 μM for MexB) and is used in microbiology research to study efflux-mediated antibiotic resistance and to potentiate antimicrobial agents in vitro. The compound is supplied as a high-purity solid suitable for biochemical and cellular assays.
  • Potent inhibitor of AcrB and MexB efflux pump subunits.
  • Measured KD values near 1.15 μM (AcrB) and 3.57 μM (MexB).
  • High reported purity suitable for biochemical assays.
  • Solid small-molecule formulation amenable to in vitro testing.
  • Useful for research on efflux-mediated antibiotic resistance and potentiation assays.
Catalog No. 50-002-39474 Supplier Medchemexpress LLC Supplier No. HY12481925MG
D13-9001 is a small-molecule inhibitor of RND-family bacterial efflux pump subunits AcrB (AcrAB-TolC) and MexB (MexAB-OprM). It shows binding affinities (KD ≈ 1.15 μM for AcrB and 3.57 μM for MexB) and is used in microbiology research to study efflux-mediated antibiotic resistance and to potentiate antimicrobial agents in vitro. The compound is supplied as a high-purity solid suitable for biochemical and cellular assays.
  • Potent inhibitor of AcrB and MexB efflux pump subunits.
  • Measured KD values near 1.15 μM (AcrB) and 3.57 μM (MexB).
  • High reported purity suitable for biochemical assays.
  • Solid small-molecule formulation amenable to in vitro testing.
  • Useful for research on efflux-mediated antibiotic resistance and potentiation assays.