Learn More
Medchemexpress LLC 2,2',4,4'-tetrahydroxybenzophenone | 131-55-5 | MFCD00002278 | 99.7% | 246.22 g/mol | C13H10O5 | 25 G
SDP
Catalog No. 5000302899
Encompass_Preferred
Benzophenone-2 is a substituted benzophenone used as a UV absorber and a research chemical for studies of xenobiotic-sensing and estrogenic pathways. It is supplied as a crystalline powder suitable for laboratory use.
  • High analytical purity (99.66%).
  • Useful UV absorber for photochemistry and formulation studies.
  • Modulates pregnane X receptor and estrogen receptor α activity in biological assays.
  • Solid powder; formula C13H10O5, molecular weight 246.22 g/mol.
  • Available in laboratory-scale pack sizes, including 25 g.
Catalog No. 50-003-02899 Supplier Medchemexpress LLC Supplier No. 25GHYW036120
Benzophenone-2 is a substituted benzophenone used as a UV absorber and a research chemical for studies of xenobiotic-sensing and estrogenic pathways. It is supplied as a crystalline powder suitable for laboratory use.
  • High analytical purity (99.66%).
  • Useful UV absorber for photochemistry and formulation studies.
  • Modulates pregnane X receptor and estrogen receptor α activity in biological assays.
  • Solid powder; formula C13H10O5, molecular weight 246.22 g/mol.
  • Available in laboratory-scale pack sizes, including 25 g.