Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2',4,4'-Tetrahydroxybenzophenone 98.0+%, TCI America™
SDP

Catalog No. T011825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T011825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T011825G Supplier TCI America Supplier No. T011825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 131-55-5
Molecular Formula C13H10O5
Molecular Weight (g/mol) 246.218
MDL Number MFCD00002278
InChI Key WXNRYSGJLQFHBR-UHFFFAOYSA-N
Synonym 2,2',4,4'-tetrahydroxybenzophenone, bis 2,4-dihydroxyphenyl methanone, benzophenone-2, uvinol d-50, uvinul d-50, methanone, bis 2,4-dihydroxyphenyl, 2,4,2',4'-tetrahydroxybenzophenone, unii-prr8k3h9vn, 2,2',4,4'-tetrehydroxybenzophenone, prr8k3h9vn
PubChem CID 8571
IUPAC Name bis(2,4-dihydroxyphenyl)methanone
SMILES C1=CC(=C(C=C1O)O)C(=O)C2=C(C=C(C=C2)O)O

Specifications

Melting Point 197°C
Quantity 25 g
Formula Weight 246.22
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′,4,4′-Tetrahydroxybenzophenone
EINECSNumber (4)-0131
RTECSNumber DJ1892000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.