Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2',4,4'-Tetramethylbenzophenone 96.0+%, TCI America™
SDP

Catalog No. T12325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T12325G 5 g
T123225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T12325G Supplier TCI America Supplier No. T12325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3478-88-4
Molecular Formula C17H18O
Molecular Weight (g/mol) 238.33
MDL Number MFCD00060094
InChI Key KFNQSAKXSGGDAA-UHFFFAOYSA-N
PubChem CID 12274022
IUPAC Name bis(2,4-dimethylphenyl)methanone
SMILES CC1=CC(C)=C(C=C1)C(=O)C1=C(C)C=C(C)C=C1

Specifications

Color Yellow
Boiling Point 188°C
Quantity 5 g
Formula Weight 238.33
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2′,4,4′-Tetramethylbenzophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.