Learn More
Medchemexpress LLC 2-[2-(4-benzhydrylpiperazin-1-yl)ethoxy]acetic acid dihydrochloride | 83881-54-3 | MFCD23704641 | 99.4% | C21H28Cl2N2O3 | 10MG
SDP
Catalog No. 5000259405
Encompass_Preferred
Deschloro Cetirizine dihydrochloride is the deschloro impurity of cetirizine supplied as the dihydrochloride salt. It is provided as a white to off-white solid and is intended for analytical testing, impurity profiling, and research use as a reference material with defined physicochemical properties.
Catalog No. 50-002-59405 Supplier Medchemexpress LLC Supplier No. HY13157910MG
Deschloro Cetirizine dihydrochloride is the deschloro impurity of cetirizine supplied as the dihydrochloride salt. It is provided as a white to off-white solid and is intended for analytical testing, impurity profiling, and research use as a reference material with defined physicochemical properties.