Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2,4-Difluorophenyl)pyridine 98.0+%, TCI America™
SDP

Catalog No. D34131G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34131G 1 g
D34135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D34131G Supplier TCI America Supplier No. D34131G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 391604-55-0
Molecular Formula C11H7F2N
Molecular Weight (g/mol) 191.181
MDL Number MFCD07787526
InChI Key SSABEFIRGJISFH-UHFFFAOYSA-N
PubChem CID 11263359
IUPAC Name 2-(2,4-difluorophenyl)pyridine
SMILES C1=CC=NC(=C1)C2=C(C=C(C=C2)F)F

Specifications

Color Yellow
Quantity 1 g
Formula Weight 191.18
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(2,4-Difluorophenyl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.