Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2,4-Dihydroxyphenyl)-4,6-bis(2,4-dimethylphenyl)-1,3,5-triazine 98.0+%, TCI America™
SDP

Catalog No. D45275G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45275G 5 g
D452725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D45275G Supplier TCI America Supplier No. D45275G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1668-53-7
Molecular Formula C25H23N3O2
Molecular Weight (g/mol) 397.478
MDL Number MFCD16251125
InChI Key AQFXADRZDKALKG-UHFFFAOYSA-N
PubChem CID 5464518
IUPAC Name 4-[2,6-bis(2,4-dimethylphenyl)-1H-1,3,5-triazin-4-ylidene]-3-hydroxycyclohexa-2,5-dien-1-one
SMILES CC1=CC(=C(C=C1)C2=NC(=C3C=CC(=O)C=C3O)N=C(N2)C4=C(C=C(C=C4)C)C)C

Specifications

Melting Point 205°C
Quantity 5 g
Formula Weight 397.48
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-(2,4-Dihydroxyphenyl)-4,6-bis(2,4-dimethylphenyl)-1,3,5-triazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.