Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2,4-Dinitrobenzyl)pyridine 98.0+%, TCI America™
SDP

Catalog No. D10955G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D10955G 5 g
D10951G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D10955G Supplier TCI America Supplier No. D10955G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1151-97-9
Molecular Formula C12H9N3O4
Molecular Weight (g/mol) 259.221
MDL Number MFCD00014632
InChI Key KKFNJVINGIUTIH-UHFFFAOYSA-N
PubChem CID 70854
IUPAC Name 2-[(2,4-dinitrophenyl)methyl]pyridine
SMILES C1=CC=NC(=C1)CC2=C(C=C(C=C2)[N+](=O)[O-])[N+](=O)[O-]

Specifications

Melting Point 90°C
Color Yellow
Quantity 5 g
Formula Weight 259.22
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(2,4-Dinitrobenzyl)pyridine
EINECSNumber (5)-3721
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.