Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2,4-Dinitrophenylthio)benzothiazole 98.0+%, TCI America™
SDP

Catalog No. D0848500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0848500G 500 g
D084825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0848500G Supplier TCI America Supplier No. D0848500G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17586-89-9
Molecular Formula C13H7N3O4S2
Molecular Weight (g/mol) 333.336
MDL Number MFCD00059704
InChI Key RCPUUVXIUIWMEE-UHFFFAOYSA-N
Synonym 2-(2,4-Dinitrophenylmercapto)benzothiazole
PubChem CID 77908
IUPAC Name 2-(2,4-dinitrophenyl)sulfanyl-1,3-benzothiazole
SMILES C1=CC=C2C(=C1)N=C(S2)SC3=C(C=C(C=C3)[N+](=O)[O-])[N+](=O)[O-]

Specifications

Melting Point 160°C
Quantity 500 g
UN Number 1325
Formula Weight 333.34
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 2-(2,4-Dinitrophenylthio)benzothiazole
EINECSNumber (5)-0263
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.