Learn More
Medchemexpress LLC 2-[2-(4-methoxy-2-pyridinyl)ethyl]-1H-imidazo[4,5-b]pyridine, dihydrochloride | 1216722-25-6 | MFCD11100187 | 99.6% | 327.21 g·mol⁻¹ | C14H16Cl2N4O | 5 MG
SDP
Catalog No. 5000240418
Encompass_Preferred
BYK 191023 dihydrochloride is a selective inducible nitric-oxide synthase (iNOS) inhibitor used in in vitro and in vivo research to probe iNOS-mediated pathways. It is supplied as a solid research chemical with high purity and is characterized by a defined molecular formula and molecular weight.
  • Selective inhibitor of inducible nitric-oxide synthase for mechanistic studies
  • High purity (≈99.6%) for reproducible results
  • Well-characterized molecular weight 327.21 g·mol⁻¹ and chemical formula C14H16Cl2N4O
  • Solid powder form for convenient handling and formulation
  • Storage recommendations available for both solid and solution states
  • Available in small research quantities suitable for assay development
Catalog No. 50-002-40418 Supplier Medchemexpress LLC Supplier No. HY1075845MG
BYK 191023 dihydrochloride is a selective inducible nitric-oxide synthase (iNOS) inhibitor used in in vitro and in vivo research to probe iNOS-mediated pathways. It is supplied as a solid research chemical with high purity and is characterized by a defined molecular formula and molecular weight.
  • Selective inhibitor of inducible nitric-oxide synthase for mechanistic studies
  • High purity (≈99.6%) for reproducible results
  • Well-characterized molecular weight 327.21 g·mol⁻¹ and chemical formula C14H16Cl2N4O
  • Solid powder form for convenient handling and formulation
  • Storage recommendations available for both solid and solution states
  • Available in small research quantities suitable for assay development