Learn More
Sigma Aldrich 2-[2-(4-Methoxyphenyl)ethyl]-1,3-dioxo-5-isoindolinecarboxylic acid
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- 2,3,5,6-tetrafluoro-4-(trifluoromethyl)phenol (perfluoro-p-cresol), an organofluorine compound, is an aryl fluorinated building block
- It is a colorless liquid having hygroscopic properties and a faintly phenolic odor
- It has been prepared by using octafluorotoluene as starting reagent
- Various physical properties (density, refractive index, freezing point and boiling point) of 2,3,5,6-tetrafluoro-4-(trifluoromethyl)phenol have been reported
Specifications
Specifications
| Molecular Formula | C18H15NO5 |
| Linear Formula | C18H15NO5 |
| MDL Number | MFCD02905116 |
| Quantity | 1 g |
| Molecular Weight (g/mol) | 325.32 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.