Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,2'-(4-Methylphenylimino)diethanol

Catalog No. 164127500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-412-7500G 500 g
16-412-7100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16-412-7500G Supplier Sigma Aldrich Supplier No. 164127500G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2,2'-(4-Methylphenylimino)diethanol

Specifications

Melting Point 49°C to 53°C (lit.)
Boiling Point 338°C to 340°C (lit.)
Molecular Formula C11H17NO2
Linear Formula CH3C6H4N(CH2CH2OH)2
MDL Number MFCD00020580
Quantity 500 g
Synonym 2,2 -(p-Tolylimino)diethanol; N,N-Bis(2-hydroxyethyl)-p-toluidine; N-(p-Tolyl)diethanolamine
Molecular Weight (g/mol) 195.26
Percent Purity 90%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.