Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2',4-Trimethoxybenzophenone 98.0+%, TCI America™
SDP

Catalog No. T20285G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T20285G 5 g
T202825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T20285G Supplier TCI America Supplier No. T20285G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 33077-87-1
Molecular Formula C16H16O4
Molecular Weight (g/mol) 272.3
MDL Number MFCD01098829
InChI Key FUGHBBQOPVKADC-UHFFFAOYSA-N
PubChem CID 3507164
IUPAC Name (2,4-dimethoxyphenyl)-(2-methoxyphenyl)methanone
SMILES COC1=CC(=C(C=C1)C(=O)C2=CC=CC=C2OC)OC

Specifications

Melting Point 56°C
Color White
Boiling Point 200°C
Quantity 5 g
Formula Weight 272.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′,4-Trimethoxybenzophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.