Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,4-Trimethyl-1,3-pentanediol 97.0+%, TCI America™
SDP

Catalog No. T048325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T048325G 25 g
T0483500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T048325G Supplier TCI America Supplier No. T048325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 144-19-4
Molecular Formula C8H18O2
Molecular Weight (g/mol) 146.23
MDL Number MFCD00004681
InChI Key JCTXKRPTIMZBJT-UHFFFAOYNA-N
Synonym 2,2,4-trimethyl-1,3-pentanediol, tmpd, 1,3-pentanediol, 2,2,4-trimethyl, tmpd alcohol, caswell no. 893, tmpd glycol, epa pesticide chemical code 041002, 2,2,4-trimethyl pentanediol-1,3, trimethyl-1,3-pentanediol, acmc-1brgn
PubChem CID 8946
IUPAC Name 2,2,4-trimethylpentane-1,3-diol
SMILES CC(C)C(O)C(C)(C)CO

Specifications

Melting Point 54°C
Color White
Boiling Point 232°C
Quantity 25 g
Formula Weight 146.23
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2,4-Trimethyl-1,3-pentanediol
EINECSNumber (2)-0240
RTECSNumber SA1400000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.